Learn More
ALK (phospho Y1586), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
The 2;5 chromosomal translocation is frequently associated with anaplastic large cell lymphomas (ALCLs). The translocation creates a fusion gene consisting of the ALK (anaplastic lymphoma kinase) gene and the nucleophosmin (NPM) gene: the 3' half of ALK, derived from chromosome 2, is fused to the 5' portion of NPM from chromosome 5. A recent study shows that the product of the NPM-ALK fusion gene is oncogenic. The deduced amino acid sequences reveal that ALK is a novel receptor protein-tyrosine kinase having a putative transmembrane domain and an extracellular domain. These sequences are absent in the product of the transforming NPM-ALK gene. ALK shows the greatest sequence similarity to LTK (leukocyte tyrosine kinase). ALK plays an important role in the development of the brain and exerts its effects on specific neurons in the nervous system. [provided by RefSeq]
Specifications
Specifications
| Antigen | ALK |
| Applications | ELISA, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic phosphopeptide of ALK. |
| Dilution | Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:4000) The optimal working dilution should be determined by the end user. |
| Formulation | In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide) |
| Gene | ALK |
| Gene Accession No. | Q9UM73 |
| Gene Alias | CD246/Ki-1/TFG/ALK |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.