Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

App Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116997 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-997 Supplier Abnova Corporation Supplier No. PAB14316
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of App.

Sequence: HMNVQNGKWESDPSGTKTC

Specifications

Antigen App
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of App.
Dilution Immunohistochemistry (1:500-1:3000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from whole serum
Gene App
Gene Alias AL024401/Abeta/Adap/Cvap/E030013M08Rik/appican/betaAPP
Gene Symbols App
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with blue carrier protein) corresponding to amino acids 44-62 of mouse App.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11820
Target Species Mouse, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.