Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Aspa/Nur7, Polyclonal,

Catalog No. ABN1698MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1698MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1698MI Supplier MilliporeSigma™ Supplier No. ABN1698
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Aspa/Nur7 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry, Western Blotting

Aspartoacylase (EC 3.5.1.15; UniProt Q8R3P0; also known as Acy-2, Aminoacylase-2, Nur-7) is encoded by the Aspa (also known as Acy2) gene (Gene ID 11484) in murine species. Aspartoacylase catalyzes the deacetylation of N-acetylaspartate (NAA) to generate free acetate in the central nervous system (CNS). Mutations in the ASPA gene cause the autosomal recessive neurodegenerative Canavan disease (CD) that results in inadequate lipid/myelin synthesis during development. Immunohistochemical staining reveals that aspartoacylase colocalizes with the oligodendrocyte marker CC1 throughout the brain, indicating its role in myelination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aspa/Nur7
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q8R3P0
Gene Symbols Aspa
Host Species Rabbit
Immunogen N-terminally his-tagged full-length recombinant mouse Aspa/Nur7.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_075602
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.