Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP7A, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89122137 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-137 Supplier Abnova Corporation Supplier No. PAB20632
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant ATP7A.

This gene encodes a transmembrane protein that functions in copper transport across membranes. The protein localizes to the trans-Golgi network, where it is predicted to supply copper to copper-dependent enzymes in the secretory pathway. The protein relocalizes to the plasma membrane under conditions of elevated extracellular copper and functions in the efflux of copper from cells. Mutations in this gene result in Menkes disease, X-linked cutis laxa, and occipital horn syndrome. [provided by RefSeq

Sequence: TETLGTCIDFQVVPGCGISCKVTNIEGLLHKNNWNIEDNNIKNASLVQIDASNEQSSTSSSMIIDAQISNALNAQQYKVLIGNREWMIRNGLVINNDVNDFMTEHERKGRTAVLVAVDDELC

Specifications

Antigen ATP7A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ATP7A.
Dilution Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene ATP7A
Gene Accession No. Q04656
Gene Alias FLJ17790/MK/MNK
Gene Symbols ATP7A
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human ATP7A.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 538
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.