Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMX (phospho Y566), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89117940 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-940 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-940 Supplier Abnova Corporation Supplier No. PAB15894
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of BMX.

This gene encodes a nonreceptor tyrosine kinase belonging to the Tec kinase family. The protein contains a PH-like domain, which mediates membrane targeting by binding to phosphatidylinositol 3,4,5-triphosphate (PIP3), and a SH2 domain that binds to tyrosine-phosphorylated proteins and functions in signal transduction. The protein is implicated in several signal transduction pathways including the Stat pathway, and regulates differentiation and tumorigenicity of several types of cancer cells. Multiple alternatively spliced variants, encoding the same protein, have been identified

Specifications

Antigen BMX
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of BMX.
Dilution ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene BMX
Gene Alias ETK/PSCTK2/PSCTK3
Gene Symbols BMX
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding Y566 of human BMX.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 660
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.