Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Bovine IgG Fc Secondary Antibody [Janelia Fluor 525], Novus Biologicals™
SDP

Catalog No. NB785277 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB785277 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB785277 Supplier Novus Biologicals Supplier No. NBP172769JF525
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IgG Fc Polyclonal antibody specifically detects IgG Fc in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen IgG Fc
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Rabbit anti-Bovine IgG Fc fragment Secondary Antibody was produced by repeated immunization with bovine IgG F(c) fragment in rabbit.
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.