Learn More
Genscript Corporation MonoRab™ Rabbit Anti-Camelid VHH Antibody [iFluor 647], mAb
Supplier:  Genscript Corporation A01994200
Encompass_Preferred
The MonoRab™ Rabbit Anti-Camelid VHH Antibody [iFluor 647], mAb is specific for Camelid VHH (Alpaca, Llama and Camel). This monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.
Catalog No. 50-227-2012
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.