Learn More
Description
Specifications
Specifications
| Antigen | IgM |
| Applications | Dot Blot, ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | HRP |
| Dilution | Western Blot 1:1000 - 1:10000, ELISA 1:10000 - 1:50000, Immunohistochemistry 1:500 - 1:2500, Immunohistochemistry-Paraffin |
| Formulation | 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 10Mg/ml BSA - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate |
| Gene Alias | AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH |
| Host Species | Rabbit |
| Immunogen | Dog IgM u mu chain |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
