Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBLN4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89119329 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-329 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-329 Supplier Abnova Corporation Supplier No. PAB17664
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CBLN4.

Cerebellin is a sixteen aa peptide found mainly in the adrenal medulla, where it has been shown to have a neuromodulatory function. Cerebellin is derived from precerebellin, a protein with sequence similarity to the noncollagen domain of complement component C1qB. The protein encoded by this gene is a glycoprotein which shares sequence similarity with precerebellin. [provided by RefSeq

Specifications

Antigen CBLN4
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CBLN4.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gene CBLN4
Gene Accession No. Q9NTU7
Gene Alias CBLNL1
Gene Symbols CBLN4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal of human CBLN4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140689
Test Specificity This antibody detects endogenous levels of total CBLN4 protein.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.