Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-CGI-58, Polyclonal,

Catalog No. ABS1616MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1616MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1616MI Supplier MilliporeSigma™ Supplier No. ABS1616
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects CGI-58 in Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

1-acylglycerol-3-phosphate O-acyltransferase ABHD5 (EC:2.3.1.51; UniProt Q9DBL9; also known as Abhydrolase domain-containing protein 5, CGI-58, Comparative gene identification-58, Lipid droplet-binding protein CGI-58) is encoded by the Abhd5 (also known as CDS, IECN5, NCIE2, 1300003D03Rik, 2010002J10Rik) gene in murine species (Gene ID 67469). The comparative gene identification-58 (CGI-58) functions as a cofactor of adipose triglyceride lipase/ATGL-mediated triacylglycerol (TG) hydrolytic activity, which is essential for efficient catabolism of cellular TG stores. Mutations in the genes coding for ATGL or CGI-58 in humans cause neutral lipid storage disease characterized by TG accumulation in multiple tissues. CGI-58 gene mutations provoke severe ichthyosis and hepatosteatosis in humans and mice. Mice lacking CGI-58 exclusively in muscle (CGI-58KOM) show TG accumulation in both skeletal and cardiac muscle tissues and develop severe cardiac steatosis and cardiomyopathy as a result of impaired TG catabolism and mitochondrial fatty acid oxidation. Immunocytochemistry analysis reveals diffuse cytoplasmic staining of CGI-58 in 3T3-L1 preadipocytes, while CGI-58 staining is found exclusively on the surface of lipid droplets in differentiated 3T3-L1 adipocytes. CGI-58 binds to lipid droplets by interacting with perilipin C-terminal sequence, while activation of cellular PKA disperses CGI-58 localization from the lipid droplets to cytosol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGI-58
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. Q9DBL9
Gene Symbols Abhd5
Host Species Rabbit
Immunogen His-tagged recombinant protein corresponding to mouse CGI-58.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_080455
Target Species Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.