Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL5A1/COL5A2/COL5A3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116711 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-711 Supplier Abnova Corporation Supplier No. PAB13507
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native COL5A1/COL5A2/COL5A3.

Specifications

Antigen COL5A1/COL5A2/COL5A3
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native COL5A1/COL5A2/COL5A3.
Dilution ELISA (>1:200) Immunohistochemistry (1:500) Immunofluorescence (1:80) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Symbols COL5A1
Host Species Rabbit
Immunogen Native purified COL5A1/COL5A2/COL5A3 from human placenta.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1289/1290/50509
Test Specificity This antibody reacts with human collagen type V 100% binding in RIA at 1:100 dilution, cross-reactivity with human collagen type I, III and IV <0.1%; human fibronectin and laminin <0.1%.
Target Species Bovine, Human, Pig
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.