Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit anti-DDDDK Tag (FLAG Tag), MP Biomedicals™
Rabbit Monoclonal Antibody
Supplier: MP Biomedicals, Inc 08L100031
This item is not returnable.
View return policy
Description
- Rabbit anti-DDDDK Tag (FLAG Tag) mAb offers high specificity and potency and consistency against FLAG Tag, which has been experimentally validated in Western blotting (WB), Immunohistochemistry (IHC), Immunoprecipitation (IP), and Immunofluorescence (IF) assays.
- FLAG-tag, or FLAG octapeptide, or FLAG epitope, is a polypeptide protein tag that can be added to a protein using recombinant DNA technology, having the sequence motif DYKDDDDK. It has been used for studying proteins in living cells and for protein purification by affinity chromatography.
- It has been used to separate recombinant, overexpressed protein from wild-type protein expressed by the host organism.
- It can also be used in the isolation of protein complexes with multiple subunits because its mild purification procedure tends not to disrupt such complexes.
- It has been used to obtain proteins of sufficient purity and quality to carry out 3D structure determination by x-ray crystallography.
- A FLAG tag can be used in many different assays that require recognition by an antibody. If there is no antibody against a given protein, adding a FLAG tag to a protein allows the protein to be studied with an antibody against the FLAG sequence.

Specifications
DDDDK Tag (FLAG Tag) | |
Monoclonal | |
In PBS buffer with 0.02% sodium azide, 0.05% BSA, 50% glycerol; pH 7.3 to 7.4 | |
Synthetic peptide corresponding to DDDDK tag | |
100 μL | |
Secondary | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Affinity Purification | |
RUO | |
Purified |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction