Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPAGT1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127348 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-348 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-348 Supplier Abnova Corporation Supplier No. PAB25725
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of DPAGT1.

The protein encoded by this gene is an enzyme that catalyzes the first step in the dolichol-linked oligosaccharide pathway for glycoprotein biosynthesis. This enzyme belongs to the glycosyltransferase family 4. This protein is an integral membrane protein of the endoplasmic reticulum. The congenital disorder of glycosylation type Ij is caused by mutation in the gene encoding this enzyme. [provided by RefSeq

Specifications

Antigen DPAGT1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of DPAGT1.
Dilution Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene DPAGT1
Gene Accession No. NP_001373
Gene Alias ALG7/CDG-Ij/D11S366/DGPT/DPAGT/DPAGT2/G1PT/GPT/UAGT/UGAT
Gene Symbols DPAGT1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids at N-terminus of human DPAGT1.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1798
Test Specificity At least four isoforms of DPAGT1 are known to exist; this antibody will recognize the two longest isoforms. DPAGT1 antibody is predicted to not cross-react with UHRF1BP1.
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.