Learn More
Description
Specifications
Specifications
| Antigen | IgA/IgG/IgM (H+L) |
| Applications | Immunoelectrophoresis |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 |
| Formulation | Stable whole antiserum with No Preservative |
| Host Species | Rabbit |
| Immunogen | Purified normal Ig fractions isolated from pooled duck serum. Freund's complete adjuvant is used in the first step of the immunization procedure. |
| Purification Method | Whole Antiserum |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
