Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP22 Rabbit anti-Bovine, Horse, Human, Monkey, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89129848 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-848 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-848 Supplier Abnova Corporation Supplier No. PAB28346
Only null left
Add to Cart
Add to Cart

Rabbit polyconal antibody raised against synthetic peptide of DUSP22.

Specifications

Antigen DUSP22
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyconal antibody raised against synthetic peptide of DUSP22.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-22 ug/ml) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene DUSP22
Gene Accession No. Q9NRW4
Gene Alias JKAP/JSP1/MKPX/VHX
Gene Symbols DUSP22
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids at N-terminus of human DUSP22.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56940
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except DUSP15 (59%).
Target Species Bovine, Equine, Human, Monkey, Rabbit
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.