Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF3K Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89119381 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-381 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-381 Supplier Abnova Corporation Supplier No. PAB17716
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of EIF3K.

The 700kDa eukaryotic translation initiation factor-3 (eIF3) is the largest eIF and contains at least 12 subunits, including EIF2S12. eIF3 plays an essential role in translation by binding directly to the 40S ribosomal subunit and promoting formation of the 40S preinitiation complex (Mayeur et al., 2003 [PubMed 14519125]).[supplied by OMIM

Specifications

Antigen EIF3K
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of EIF3K.
Dilution Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gene EIF3K
Gene Accession No. Q9UBQ5
Gene Alias ARG134/EIF3-p28/EIF3S12/HSPC029/M9/MSTP001/PLAC-24/PLAC24/PRO1474/PTD001
Gene Symbols EIF3K
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal of human EIF3K.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27335
Test Specificity This antibody detects endogenous levels of total EIF3K protein.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.