Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-ERG, Clone: EPR3864, Abnova™

Catalog No. 89108333 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-333 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-333 Supplier Abnova Corporation Supplier No. MAB10018

Specifications

Antigen ERG
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone EPR3864
Conjugate Unconjugated
Formulation In PBS (BSA, 0.09% sodium azide)
Gene Symbols ERG
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human ERG.
Purification Method Protein A purification
Quantity 500 μL
Primary or Secondary Primary
Gene ID (Entrez) 2078
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C is stable until expiration date.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.