Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Feline IgG Fc Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
SDP

Catalog No. NBP172645 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP172645 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP172645 Supplier Novus Biologicals Supplier No. NBP172645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Feline samples. It is validated for Dot Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IgG Fc
Applications Dot Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Alkaline Phosphatase (AP)
Dilution Western Blot 1:500 - 1:2500, ELISA 1:4000 - 1:16000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:10-1:500, Dot Blot
Host Species Rabbit
Immunogen Cat IgG F(c) fragment
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity No reaction was observed against Cat IgG F(ab).
Target Species Feline
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.