Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-GFP-Tag, MP Biomedicals™

Catalog No. MP08L100036 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08L100036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP08L100036 Supplier MP Biomedicals, Inc Supplier No. 08L100036
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

  • Rabbit anti GFP-Tag mAb offers high specificity and potency and consistency against the GFP tag, which has been experimentally validated in Western blotting (WB) and Immunofluorescence (IF) assays.
  • The green fluorescent protein (GFP) is a protein composed of 238 amino acid residues (26.9 kDa) that exhibit bright green fluorescence when exposed to light in the blue to ultraviolet range.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFP-Tag
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Concentration WB: 1:500 - 1:2000; IF: 1:50 - 1:200
Conjugate Unconjugated
Formulation In PBS buffer with 0.02% sodium azide, 0.05% BSA, 50% glycerol; pH 7.3 to 7.4
Host Species Rabbit
Immunogen Full-length recombinant GFP protein
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Secondary
Form Purified
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.