Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLRA1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121326 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-326 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-326 Supplier Abnova Corporation Supplier No. PAB19797
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GLRA1.

The protein encoded by this gene is a subunit of a pentameric inhibitory glycine receptor. The receptor mediates postsynaptic inhibition in the central nervous system. Defects in this gene are a cause of startle disease (STHE), also known as hereditary hyperekplexia or congenital stiff-person syndrome. Two transcript variants encoding different isoforms have been found for this gene

Specifications

Antigen GLRA1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GLRA1.
Dilution ELISA (1:16000-1:60000) Western Blot (1:200-1000) The optimal working dilution should be determined by the end user.
Formulation In serum (0.02% sodium azide)
Gene GLRA1
Gene Accession No. P23415
Gene Alias MGC138878/MGC138879/STHE
Gene Symbols GLRA1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 16 amino acids near N-terminus of human GLRA1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2741
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.