Learn More
Rabbit anti-Goat IgG F(ab')2 Secondary Antibody (Pre-adsorbed), R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG F(ab')2 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000 |
| Gene Alias | immunoglobulin heavy constant gamma 1 (G1m marker) |
| Host Species | Rabbit |
| Immunogen | Goat IgG F(ab')2 fragment |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.