Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit anti-Goat IgG, Whole Molecule Polyclonal Secondary Antibody, HRP, MP Biomedicals
Supplier: MP Biomedicals, Inc 0855363
Description
Host Species: Rabbit
Species Reactivity: Goat

Specifications
IgG, Whole Molecule | |
HRP | |
2 mL | |
Goat |
Polyclonal | |
Rabbit | |
Secondary | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction