Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Guinea Pig IgG Fc Secondary Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB483993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB483993 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB483993 Supplier Novus Biologicals Supplier No. NBP172735PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Guinea Pig samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG Fc
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Host Species Rabbit
Immunogen Guinea Pig IgG F(c) fragment
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Guinea Pig
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.