Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Human IgG (H+L) Secondary Antibody [Janelia Fluor 525], R&D Systems™
SDP

Catalog No. NB786095 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB786095 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB786095 Supplier R&D Systems Supplier No. NBP175070JF525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay

Specifications

Antigen IgG (H+L)
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay (CIA)
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen This Rabbit anti-Human IgG (H+L) Secondary Antibody was developed against purified human IgG (H&L).
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.