Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Human IgG Heavy Chain Secondary Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB543464 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB543464 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB543464 Supplier Novus Biologicals Supplier No. NBP222138APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IgG Heavy Chain Polyclonal specifically detects IgG Heavy Chain in Human samples. It is validated for Dot Blot, ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG Heavy Chain
Applications Dot Blot, ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Rabbit
Immunogen Human IgG gamma heavy chain
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.