Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Human IgG1 Fc Secondary Antibody (245), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP340578V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP340578V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP340578V Supplier Novus Biologicals Supplier No. NBP340578V
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IgG1 Fc Monoclonal specifically detects IgG1 Fc in Human samples. It is validated for ELISA.

Specifications

Antigen IgG1 Fc
Applications ELISA
Classification Monoclonal
Clone 245
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IgG1 Fc (Uniprot#: P01857-1; Glu99-Lys330,C103S). HEK293 used as expression system.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.