Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit anti-Llama IgG (H+L) Secondary Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130113
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Llama samples. It is validated for Western Blot.Specifications
| IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Llama | |
| IgG |
| Western Blot | |
| 1.78 mg/mL | |
| Western Blot 1:30000 | |
| Purified Llama IgG | |
| 0.25 mg | |
| Adaptive Immunity, Immunology | |
| This rabbit anti-llama secondary antibody recognizes both heavy and light chains | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction