Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Llama IgG Heavy Chain Secondary Antibody, Biotin, R&D Systems™
SDP

Catalog No. NBP147632 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147632 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147632 Supplier R&D Systems Supplier No. NBP147632

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

IgG Heavy Chain Polyclonal specifically detects IgG Heavy Chain in Llama samples. It is validated for Western Blot.

Specifications

Antigen IgG Heavy Chain
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Rabbit
Immunogen Llama IgG Heavy Chain
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity Llama.
Target Species Llama
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.