Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRN4 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89127713 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-713 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-713 Supplier Abnova Corporation Supplier No. PAB26073
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of LRRN4.

Specifications

Antigen LRRN4
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of LRRN4.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene LRRN4
Gene Alias C20orf75/FLJ23994/MGC25027/NLRR4/dJ1056H1.1
Gene Symbols LRRN4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 16 amino acids at N-terminus of human LRRN4.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164312
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except SH2B1 (44%).
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.