Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2K1/MAP2K2 Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116038 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-038 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-038 Supplier Abnova Corporation Supplier No. PAB12707
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of MAP2K1/MAP2K2.

Specifications

Antigen MAP2K1/MAP2K2
Applications Dot Blot, ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of MAP2K1/MAP2K2.
Dilution Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 7.2 (BSA, 10% Proclin300)
Gene Alias MAPKK1/MEK1/MKK1/PRKMK1
Gene Symbols MAP2K1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues surrounding S218 and S222 of human MAP2K1/MAP2K2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5604/5605
Test Specificity This antibody recognizes ∼44, 45 KDa of human MAP2K1 without phosphorylation sites of Ser 218/222 or MAP2K2 (pS222/226). It does not cross react to phospho MAP2K1/MAP2K2.
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.