Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP3K7IP2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89128834 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-834 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-834 Supplier Abnova Corporation Supplier No. PAB27182
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of MAP3K7IP2.

The protein encoded by this gene is an activator of MAP3K7/TAK1, which is required for for the IL-1 induced activation of nuclear factor kappaB and MAPK8/JNK. This protein forms a kinase complex with TRAF6, MAP3K7 and TAB1, thus serves as an adaptor linking MAP3K7 and TRAF6. This protein, TAB1, and MAP3K7 also participate in the signal transduction induced by TNFSF11/RANKl through the activation of the receptor activator of NF-kappB (TNFRSF11A/RANK), which may regulate the development and function of osteoclasts. [provided by RefSeq

Specifications

Antigen MAP3K7IP2
Applications ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of MAP3K7IP2.
Dilution Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene MAP3K7IP2
Gene Alias FLJ21885/KIAA0733/TAB2
Gene Symbols MAP3K7IP2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 14 amino acids at C-terminus of human MAP3K7IP2.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23118
Test Specificity MAP3K7IP2 antibody is human specific. MAP3K7IP2 antibody is predicted not to cross-react with other TAB proteins.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.