Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Mouse IgA Antibody, HRP, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP173722 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP173722 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP173722 Supplier Bio-techne Sales Corp Supplier No. NBP173722

Specifications

Antigen IgA
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate HRP
Dilution Western Blot 1:2000 - 1:10000, ELISA 1:200000, Immunohistochemistry 1:1000 - 1:5000, Immunohistochemistry-Paraffin
Gene Alias Immunoglobulin A
Host Species Rabbit
Immunogen Mouse IgA heavy chain
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3493
Test Specificity This product was prepared from monospecific antiserum by immunoaffinity chromatography using Mouse IgA coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Rabbit Serum, Mouse IgA, and Mouse Serum. Specificity was confirmed by ELISA at less than 1% cross reactivity against other mouse heavy and light chain isotypes.
Reconstitution Reconstitute with 0.1 ml deionized water.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgA
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.