Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit F(ab')2 anti-Mouse IgG (H+L) Secondary Antibody, PE (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NB1207000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1207000 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1207000 Supplier Novus Biologicals Supplier No. NB1207000
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Dot Blot.

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate PE
Dilution Flow Cytometry 1:100 - 1:250, Immunocytochemistry/Immunofluorescence 1:100 - 1:250
Host Species Rabbit
Immunogen Rabbit F(ab')2 anti-Mouse IgG (H+L) Secondary Antibody [PE] (Pre-adsorbed) was produced by repeated immunization with Mouse IgG whole molecule in rabbit.
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Coupling to R-PE was followed by size exclusion chromatography to purify conjugate from unreacted R-PE and antibody. No reaction was observed against anti-Pepsin, anti-Rabbit IgG F(c) or Human Serum Proteins.
Target Species Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.