Learn More
Rabbit anti-Mouse IgG Fc Secondary Antibody, Alkaline Phosphatase, R&D Systems™

Description
Specifications
Specifications
| Antigen | IgG Fc |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Alkaline Phosphatase (AP) |
| Dilution | Western Blot 1:500 - 1:2500, ELISA 1:5000 - 1:15000, Immunohistochemistry 1:200 - 1:1000 |
| Host Species | Rabbit |
| Immunogen | Rabbit anti-Mouse IgG Fc fragment Secondary Antibody was produced by repeated immunization with mouse IgG F(c) fragment in rabbit. |
| Purification Method | Affinity Purified |
| Quantity | 1 mg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.