Learn More
Description
Specifications
Specifications
| Antigen | IgG Fc |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000 - 1:10000, ELISA 1:100000 - 1:250000, Immunohistochemistry 1:1000 - 1:5000 |
| Formulation | 0.05M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0 with 10 mg/ml BSA- Immunoglobulin and Protease free with 0.01% Sodium Azide |
| Host Species | Rabbit |
| Immunogen | Rabbit anti-Mouse IgG Fc fragment Secondary Antibody was produced by repeated immunization with mouse IgG F(c) fragment in rabbit. |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
