Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Mouse IgG (H+L) Secondary Antibody [Janelia Fluor 525], Novus Biologicals™
SDP

Catalog No. NB785213 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB785213 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB785213 Supplier Novus Biologicals Supplier No. NBP172731JF525
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay

Specifications

Antigen IgG (H+L)
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Fluorophore-Linked Immunosorbent Assay
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Rabbit anti-Mouse IgG (H+L) Secondary Antibody (Pre-adsorbed) was produced by repeated immunization with mouse IgG whole molecule in rabbit
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.