Learn More
Rabbit anti-Mouse IgG (H+L) Secondary Antibody [Janelia Fluor 669], R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Fluorophore-Linked Immunosorbent Assay |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Rabbit anti-Mouse IgG (H+L) Secondary Antibody (Pre-adsorbed) was produced by repeated immunization with mouse IgG whole molecule in rabbit |
| Purification Method | Multi-step |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.