Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit anti-Mouse IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™

Rabbit Polyclonal Antibody
$219.50
Specifications
| Antigen | IgG (H+L) |
|---|---|
| Concentration | 4.5mg/mL |
| Dilution | Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 |
| Applications | Flow Cytometry, Immunomicroscopy |
| Classification | Polyclonal |
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunomicroscopy.Specifications
| IgG (H+L) | |
| Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 | |
| Polyclonal | |
| Rabbit | |
| IgG | |
| Affinity purified |
| 4.5mg/mL | |
| Flow Cytometry, Immunomicroscopy | |
| Unconjugated | |
| Adaptive Immunity, Immunology | |
| Secondary | |
| Based on IEP, this antibody reacts with heavy gamma chains on mouse IgG and light chains on all mouse immunoglobulins. This antibody has been pre-adsorbed against human serum proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title