Learn More
Rabbit anti-Mouse IgG1 Heavy Chain Secondary Antibody, DyLight 800, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG1 Heavy Chain |
| Applications | Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | DyLight 800 |
| Dilution | Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:5000, Fluorophore-linked immunosorbent assay 1:20000 |
| Host Species | Rabbit |
| Immunogen | Mouse IgG1 heavy chain |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.