Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Mouse IgG1 Secondary Antibody (20) [DyLight 680], Novus Biologicals™
SDP

Catalog No. NBP343768FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP343768FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP343768FR Supplier Novus Biologicals Supplier No. NBP343768FR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IgG1 Monoclonal specifically detects IgG1 in Mouse samples. It is validated for ELISA.

Specifications

Antigen IgG1
Applications ELISA
Classification Monoclonal
Clone 20
Conjugate DyLight 680
Dilution ELISA
Formulation 50 mM Sodium Borate
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Host Species Rabbit
Immunogen Recombinant Mouse IgG1 Fc Protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.