Learn More
Rabbit anti-Mouse IgG3 Heavy Chain Secondary Antibody, DyLight 680, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG3 |
| Applications | ELISA, Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809 |
| Gene Symbols | IGHG3 |
| Host Species | Rabbit |
| Immunogen | Mouse IgG3 heavy chain |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.