Learn More
Rabbit anti-Mouse Lambda Light Chain Secondary Antibody, FITC, R&D Systems™
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Antigen | Lambda Light Chain |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Fluorescein |
| Dilution | Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 |
| Gene Alias | IGLC, immunoglobulin lambda constant group |
| Gene Symbols | IGLC1 |
| Host Species | Rabbit |
| Immunogen | Mouse lambda free chain |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.