Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE3A Rabbit anti-Horse, Human, Monkey, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89118317 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-317 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-317 Supplier Abnova Corporation Supplier No. PAB16445
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PDE3A.

Specifications

Antigen PDE3A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PDE3A.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene PDE3A
Gene Alias CGI-PDE
Gene Symbols PDE3A
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human PDE3A.
Purification Method Immunoaffinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5139
Test Specificity Internal domain of human.
Target Species Equine, Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.