Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLCE1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89122319 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-319 Supplier Abnova Corporation Supplier No. PAB20821

Rabbit polyclonal antibody raised against recombinant PLCE1.

This gene encodes a phospholipase enzyme that catalyzes the hydrolysis of phosphatidylinositol-4,5-bisphosphate to generate two second messengers: inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG). These second messengers subsequently regulate various processes affecting cell growth, differentiation, and gene expression. This enzyme is regulated by small monomeric GTPases of the Ras and Rho families and by heterotrimeric G proteins. In addition to its phospholipase C catalytic activity, this enzyme has an N-terminal domain with guanine nucleotide exchange (GEF) activity. Mutations in this gene cause early-onset nephrotic syndrome; characterized by proteinuria, edema, and diffuse mesangial sclerosis or focal and segmental glomerulosclerosis. Alternative splicing results in multiple transcript variants encoding distinct isoforms

Sequence: NSQEETLEFVADYSGQDNFLQRVGQNGLKNSEKESTVNSIFQVIRSCNRSLETDEEDSPSEGNSSRKSSLKDKSRWQFIIGDLLDSDNDIFEQSKEYDSHGSEDSQKAFDHGTELIPWYVLSIQADVHQFLLQGATVIHYDQD

Specifications

Antigen PLCE1
Applications Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description phospholipase C, epsilon 1
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene PLCE1
Gene Alias FLJ23659, KIAA1516, MGC167842, NPHS3, PLCE
Gene Symbols PLCE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human PLCE1.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51196
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.