Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Postn Rabbit anti-Birds, Chicken, Human, Mammals, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89118745 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-745 Supplier Abnova Corporation Supplier No. PAB16941
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Postn.

Specifications

Antigen Postn
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Postn.
Dilution Western Blot (1:1000) Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene Postn
Gene Alias AI747096/OSF-2/Osf2/PN/Periostin/peri
Gene Symbols Postn
Host Species Rabbit
Immunogen Recombinant protein corresponding to 188 amino acids from the C-terminus of mouse Postn.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50706
Test Specificity This antibody has been directly tested for reactivity in chicken, human, mouse and rat. This antibody is expected to work on all mammalian and avian species. specific to the ∼93 KDa periostin doublet in mouse lung extract. Consistent with the fact that this antibody is made against an alternatively spliced region of periostin, this antibody recognizes only the largest two of the three forms of periostin recognized on Western blots by the Pan periostin antibody and also shows a distinctive staining pattern by immunohistochemistry. This antibody works well for immunohistochemistry on paraformaldehyde-fixed sections with a simple antigen-retrieval protocol (incubate slides for 20 minutes at 90 °C in 10 mM sodium citrate (pH 6.0)/ 0.1 % Tween-20).
Target Species Avian, Chicken, Human, Mammalia, Mouse, Rat
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.