Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGDS Rabbit anti-Bovine, Cat, Horse, Human, Mouse, Pig, Primates, Rabbit, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115563 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-563 Supplier Abnova Corporation Supplier No. PAB12054
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant PTGDS.

The protein encoded by this gene is a glutathione-independent prostaglandin D synthase that catalyzes the conversion of prostaglandin H2 (PGH2) to postaglandin D2 (PGD2). PGD2 functions as a neuromodulator as well as a trophic factor in the central nervous system. PGD2 is also involved in smooth muscle contraction/relaxation and is a potent inhibitor of platelet aggregation. This gene is preferentially expressed in brain. Studies with transgenic mice overexpressing this gene suggest that this gene may be also involved in the regulation of non-rapid eye movement sleep. [provided by RefSeq

Specifications

Antigen PTGDS
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant PTGDS.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (30% glycerol, 0.09% sodium azide)
Gene PTGDS
Gene Alias LPGDS/PDS/PGD2/PGDS/PGDS2
Gene Symbols PTGDS
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-50 of human PTGDS.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5730
Test Specificity The epitope recognized by this antibody maps to region within residue 1-50 of human PTGDS. [Swiss-Prot# P41222]
Target Species Bovine, Equine, Feline, Human, Mouse, Pig, Primate, Rabbit, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.