Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYDC1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114142 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-142 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-142 Supplier Abnova Corporation Supplier No. PAB0326
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant PYDC1.

Specifications

Antigen PYDC1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant PYDC1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene PYDC1
Gene Alias ASC2/POP1/PYC1
Gene Symbols PYDC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length human PYDC1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 260434
Target Species Gerbil, Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.