Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDK2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89122261 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-261 Supplier Abnova Corporation Supplier No. PAB20760

Rabbit polyclonal antibody raised against recombinant SDK2.

The protein encoded by this gene is a member of the immunoglobulin superfamily. The protein contains two immunoglobulin domains and thirteen fibronectin type III domains. Fibronectin type III domains are present in both extracellular and intracellular proteins and tandem repeats are known to contain binding sites for DNA, heparin and the cell surface. This protein, and a homologous mouse sequence, are very similar to the Drosophila sidekick gene product but the specific function of this superfamily member is not yet known. Evidence for alternative splicing at this gene locus has been observed but the full-length nature of additional variants has not yet been determined. [provided by RefSeq

Sequence: SKKYAKKTDSGNSAKSGALGHSEMMSLDESSFPALELNNRRLSVKNSFCRKNGLYTRSPPRPSPGSLHYSDEDVTKYNDLIPAESSSLTEKPSEISDSQ

Specifications

Antigen SDK2
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description sidekick homolog 2 (chicken)
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene SDK2
Gene Alias FLJ10832, KIAA1514
Gene Symbols SDK2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human SDK2.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54549
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.