Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA3B Rabbit anti-Bovine, Dog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115549 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-549 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-549 Supplier Abnova Corporation Supplier No. PAB12040
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SEMA3B.

The semaphorin/collapsin family of molecules plays a critical role in the guidance of growth cones during neuronal development. The secreted protein encoded by this gene family member is important in axonal guidance and has been shown to act as a tumor suppressor by inducing apoptosis. [provided by RefSeq

Specifications

Antigen SEMA3B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SEMA3B.
Dilution Western Blot (1:5000-1:20000) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene SEMA3B
Gene Alias FLJ34863/LUCA-1/SEMA5/SEMAA/SemA/semaV
Gene Symbols SEMA3B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 100-200 of human SEMA3B.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7869
Test Specificity This antibody is specific to SEMA 3B.
Target Species Bovine, Canine, Human, Mouse, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.