Learn More
Bioworld Rabbit Anti-Sheep IgG Antibody (Polyclonal IgG) - HRP, 200 μg
Supplier:  Bioworld 203300131
Encompass_Preferred
Rabbit Anti-Sheep IgG Antibody (Polyclonal IgG) - HRPCat#20330013DetailsHost:RabbitIsotype:IgGClass:PolyclonalImmunogen:Sheep IgGConjugate:HRPForm InformationForm:LiquidConcentration:2 mg/mLPurification:Protein A ChromatrographyStorage Buffer:PBS with 0.1% BSAPreservative:Upon Request (0.02% Sodium Azide)Storage:Shipped at 4°C. Upon delivery aliquot and store at -20°C. Avoid freeze / thaw cycles.Usage RecommendationsWestern Blot (WB):1:3000-1:5000ELISA:1:150-1:400Immunoprecipitation (IP):1:350 - 1:500
Catalog No. 50-198-354
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.